Iron ICP Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1021-100ML E1106-100ML E1148-100ML E1218-100ML
HONCE1021-100MLEA 91.53 CHF
HONCE1021-100ML HONCE1106-100ML HONCE1148-100ML HONCE1218-100ML
Iron ICP Standards, (CRMs), Fluka™
Standards Kationischer Standards Eisen-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Zertifizierungen: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).