Cesium ICP standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1016-100ML E1105-100ML E1143-100ML E1213-100ML
HONCE1016-100MLEA 91.53 CHF
HONCE1016-100ML HONCE1105-100ML HONCE1143-100ML HONCE1213-100ML
Cesium ICP standards, (CRMs), Fluka™
Standards Kationischer Standards Cäsium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Zertifizierungen: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).