Diethylether stabilisiert ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™

Lieferant: Honeywell Chemicals
Gefahr

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

65756-195L
HONC65756-195LEA 3066.76 CHF
HONC65756-195L
Diethylether stabilisiert ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Diethylether
Formel: (CH₃CH₂)₂O
MW: 74.12 g/mol
Siedepunkt: 34…35 °C
Melting Pt: -123
Dichte: 0.71 g/cm3
Flash-Pt: –45 °C
MDL Number: MFCD00011646
CAS-Nummer: 60-29-7
UN: 1155
ADR: 3 I
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Assay (GC) Min. 99.8 %
Boiling range 34 - 35 °C
Density (D 20/4) 0.713 - 0.714
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.03 %
Free acid (as CH₃COOH) Max. 0.0002 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.000005 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000005 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.000005 %
Acetone (GC) Max. 0.005 %
Aldehydes complying
Carbonyl compounds (as CO) Max. 0.001 %
Ethanol (GC) Max. 0.02 %
Methanol (GC) Max. 0.02 %
Peroxides (as H₂O₂) Max. 0.00003 %
Reaction against H₂SO₄ complying
APHA Max. 10
Assay BHT (GC) 5 - 8 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR