UV-Visible/NIR Spectrophotometer, UH5700
Lieferant: VWR Collection
2J3-0013
HITA2J3-0013EA 29560 CHF
HITA2J3-0013
UV-Visible/NIR Spectrophotometer, UH5700
Spektrophotometer und Photometer UV/Vis-Spektralphotometer
The UH5700 UV-Vis-NIR Spectrophotometer is a high-precision benchtop spectrophotometer instrument designed for handling the ultraviolet, visible and near-infrared regions (190 to 3300 nm).
  • Continuously variable slit
  • Low stray light/high photometric range
  • Supports high-speed scanning
  • New software UV solutions plus
  • Multiple accessories for a wide range of measurements
UH5700 achieves best-in-class levels of low stray light and high photometric range through use of a Czerny-Turner mounted single-monochromator bright spectrometer and a newly developed