Columns for large biomolecule analysis, CIMmultus™
Lieferant: HICHROM
311.6157-6 311.5114-1.3 311.5113-2 311.5113-1.3 311.8140-2 311.6157-2 411.8136-2 411.5114-1.3 411.5113-2 411.8140-2 411.8140-1.3 411.6157-2
HISM311.6157-6EA 0 CHF
HISM311.6157-6 HISM311.5114-1.3 HISM311.5113-2 HISM311.5113-1.3 HISM311.8140-2 HISM311.6157-2 HISM411.8136-2 HISM411.5114-1.3 HISM411.5113-2 HISM411.8140-2 HISM411.8140-1.3 HISM411.6157-2
Columns for large biomolecule analysis, CIMmultus™
Chromatographiesäulen
CIMmultus™ columns are based on an innovative monolithic technology. Distinguished by large pores/flow through channels these columns are designed for the production, purification, and analytics of large proteins (IgG, IgM, PEGylated proteins), viruses (AAV, adenovirus, lentivirus, influenza, bacteriophages), VLPs, exosomes and pDNA for example.
  • Rapid, high resolution analysis
  • Flow independent separation
  • High sensitivity and reproducibility
  • Operation at high volumetric flow rates
  • Larger scale columns available and in cGMP compliant formats