Flame photometric detector (FPD) supplies
Lieferant: AGILENT HEWLETT PACKARD
19256-80045 5188-5245 5188-5953 19256-20690 1000-1438 1000-1437 19256-20910
HEWL19256-80045EA 115 CHF
HEWL19256-80045 HEWL5188-5245 HEWL5188-5953 HEWL19256-20690 HEWL1000-1438 HEWL1000-1437 HEWL19256-20910
Flame photometric detector (FPD) supplies
Chromatographie-Zubehör Chromatographie-Ersatzteile
In 2005, Agilent released an improved FPD with minimum detectable levels (MDL) of 3,6 pg/s for sulphur and 60 fg/s for phosphorus. This is more than a five times improvement for sulphur.
  • The FPD uses three gases: air and hydrogen to support the flame, and nitrogen makeup for capillary columns. The flow rates are critical for optimising performance.
The updated design is based on a one-piece, deactivated transferline jet assembly and improved optics.
Bestellinformation: Upgrade kits are available.
Order Now
TECHNISCHE DATEN
Environmentally Preferable