Box top Parafilm® dispenser
Lieferant: HEATHROW
120758
HEAT120758EA 73.5 CHF
HEAT120758
Box top Parafilm® dispenser
Spender für Verschlussfolien
Easy application, simply place the dispenser on top of the Parafilm® M box and pull the sealing film through, there is no need to remove rolls from the packaging.
  • Accommodates two 50 mm (2"), or one 100 mm (4") Parafilm® M rolls
  • Built in safety blade slides and cuts with no danger to fingertips and eliminates the need for scissors
  • Made from ABS plastic making the unit easy to clean
Lieferumfang: Supplied with two safety blades, additional replacement blades are available for purchase.