Baths and circulators, stainless steel, SAHARA PPO S21P series
Lieferant: THERMO ELECTRON LED
1563211 1553211 1523211 1533211
HAKA1563211EA 3810 CHF
HAKA1563211 HAKA1553211 HAKA1523211 HAKA1533211
Baths and circulators, stainless steel, SAHARA PPO S21P series
Umwälzthermostate Offene Umwälzsysteme Bäder mit abnehmbarem Umwälzthermostat
These heated bath circulators maintains the ambient temperature. The SAHARA series of heated bath circulators are available with a wide variety of work area dimensions to meet internal or external circulation requirements.
  • A selection of different controllers will create the perfect fit for application
  • Rugged and corrosion-resistant for high temperature applications up to 200 °C
  • Modified polyphenylene oxide baths are thermally resistant up to 100 °C
  • Deliver exceptional temperature performance with operational savings
  • Recommended for calibration,
More Product Information