1-Hexyl-3-methylimidazoliumiodid 98

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 1-hexyl-3-methylimidazolium iodide

H59487.06 H59487.18
H59487.06EA 83.7 CHF
H59487.06 H59487.18
1-Hexyl-3-methylimidazoliumiodid 98
1-Hexyl-3-methylimidazolium iodide
Formel: C₁₀H₁₉IN₂
MW: 294.18 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD12964976
CAS-Nummer: 178631-05-5

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR