1-Oxa-6-azaspiro[3.4]octane oxalate 96%
Lieferant: Thermo Fisher Scientific
Achtung
H57452.MDEA 1310 CHF
H57452.MD H57452.ME
1-Oxa-6-azaspiro[3.4]octane oxalate 96%
1-Oxa-6-azaspiro[3.4]octane oxalate
Purity: 96% Molecular Formula: C6H11NO.C2H2O4 Formula Weight: 203.19 Storage Temperature: -30°C to -10°C MDL No.: MFCD18325163
Order Now
TECHNISCHE DATEN
Environmentally Preferable