3-Brom-N-methylanilin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H55464.06
H55464.06EA 302 CHF
H55464.06
3-Brom-N-methylanilin ≥97%
3-Brom-N-methylanilin
Formel: C₇H₈BrN
MW: 186.05 g/mol
Siedepunkt: 253 °C
Dichte: 1.461
Flash-Pt: >110 °C (230 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD05664376
CAS-Nummer: 66584-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR