Propargylamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Monopropagyl amine, Prop-2-ynylamine, 3-Aminopropyne, Prop-2-yn-1-amine

H53495.06 H53495.14
H53495.06EA 125 CHF
H53495.06 H53495.14
Propargylamin 98%
Propargylamin

Purity: 98% CAS No.: 2450-71-7 Molecular Formula: C3H5N Formula Weight: 55.08 Storage Temperature: 2°C to 8°C Flash Point: 6°(43°F) Refractive Index: 1.449 Boiling Point: 83° Beilstein Reference: 773681 UN No.: 3286 MDL No.: MFCD00008198

Formel: C₃H₅N
MW: 55.08 g/mol
Siedepunkt: 83 °C
Dichte: 0.86
Flash-Pt: 6 °C (43 °F)
MDL Number: MFCD00008198
CAS-Nummer: 2450-71-7
EINECS: 219-513-8
UN: 3286
ADR: 3 II

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR