tert-Butyl propargyl ether 98%

Lieferant: Thermo Fisher Scientific
Gefahr

H53480.06 H53480.14 H53480.22
H53480.06EA 81.6 CHF
H53480.06 H53480.14 H53480.22
tert-Butyl propargyl ether 98%
tert-Butyl propargyl ether

Purity: 98% CAS No.: 16314-18-4 Molecular Formula: C7H12O Formula Weight: 112.17 Flash Point: 20°(68°F) Refractive Index: 1.418 Boiling Point: 119° UN No.: UN1993 MDL No.: MFCD07368651

Formel: C₇H₁₂O
MW: 112.172 g/mol
Siedepunkt: 119 °C
Dichte: 0.835
Flash-Pt: 20 °C (68 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD07368651
CAS-Nummer: 16314-18-4
ADR: 3 II

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR