(S)-1-(Boc-amino)-2-propanol 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: tert-Butyl (S)-(2-hydroxypropyl)carbamate

H52826.06 H52826.14
H52826.06EA 112 CHF
H52826.06 H52826.14
(S)-1-(Boc-amino)-2-propanol 97%
(S)-1-(Boc-amino)-2-propanol

Purity: 97% CAS No.: 167938-56-9 Molecular Formula: C8H17NO3 Formula Weight: 175.23 MDL No.: MFCD04974340

Formel: C₈H₁₇NO₃
MW: 175.23 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04974340
CAS-Nummer: 167938-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR