1,3-Dibromaceton 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: 1,3-Dibromopropan-2-one

H37325.14 H37325.06
H37325.14EA 595 CHF
H37325.14 H37325.06
1,3-Dibromaceton 98%
1,3-Dibromaceton

Purity: 98% CAS No.: 816-39-7 Molecular Formula: C3H4Br2O Formula Weight: 215.87 Storage Temperature: 2°C to 8°C Flash Point: 102°(216°F) Physical Form: Crystals or fused solid Sensitivity: Moisture Sensitive Appearance: White to pale yellow Melting Point: 29-30° Boiling Point: 97-98°/21mm Beilstein Reference: 1740389 UN No.: UN1759 MDL No.: MFCD00013540

Formel: BrH₂CC(O)CH₂Br
MW: 215.87 g/mol
Siedepunkt: 97…98 °C (21 mmHg)
Melting Pt: 29…30 °C
Flash-Pt: 102 °C (216 °F)
MDL Number: MFCD00013540
CAS-Nummer: 816-39-7
EINECS: 212-430-8
UN: 1759
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR