6-Fluorchinolin 97

Lieferant: Thermo Fisher Scientific
Achtung

H35900.MD H35900.03
H35900.MDEA 89.2 CHF
H35900.MD H35900.03
6-Fluorchinolin 97
6-Fluoroquinoline
Formel: C₉H₆FN
MW: 147.15 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01685512
CAS-Nummer: 396-30-5

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR