3-Amino-1H-indazol 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: 1H-Indazol-3-amine, Indazol-3-amine, 1H-Indazol-3-ylamine, Indazol-3-ylamine, 3-Amino-1H-indazole

H33435.03 H33435.09
H33435.03EA 114 CHF
H33435.03 H33435.09
3-Amino-1H-indazol 97%
3-Amino-1H-indazole
Formel: C₇H₇N₃
MW: 133.15 g/mol
Melting Pt: 150…152 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00182045
CAS-Nummer: 874-05-5
UN: 2811
ADR: 6.1 III

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR