5-Brom-2-isopropoxypyridin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H32564.03 H32564.06
H32564.03EA 166 CHF
H32564.03 H32564.06
5-Brom-2-isopropoxypyridin ≥97%
5-Brom-2-isopropoxypyridin

Purity: 97% CAS No.: 870521-31-6 Molecular Formula: C8H10BrNO Formula Weight: 216.08 Refractive Index: 1.5305

Formel: C₈H₁₀BrNO
Lager Temperatur: Raumtemperatur
MDL Number: MFCD07368869
CAS-Nummer: 870521-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR