Oxalyldichlorid 2 M in Dichlormethan

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Oxalyl chloride

H31617.AE H31617.AP
H31617.AEEA 95 CHF
H31617.AE H31617.AP
Oxalyldichlorid 2 M in Dichlormethan
Oxalyldichlorid

CAS No.: 79-37-8 Molecular Formula: C2Cl2O2 Formula Weight: 126.93 UN No.: 2922 MDL No.: MFCD00000704

Formel: C₂Cl₂O₂
Dichte: 1.335
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000704
CAS-Nummer: 79-37-8
EINECS: 201-200-2
UN: 2922
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR