trans-4-Methylcyclohexancarbonsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: trans-4-Methyl-1-cyclohexanecarboxylic acid

H27455.03 H27455.06
H27455.03EA 84.7 CHF
H27455.03 H27455.06
trans-4-Methylcyclohexancarbonsäure ≥98%
trans-4-Methylcyclohexanecarboxylic acid

Purity: 98% CAS No.: 13064-83-0 Molecular Formula: C8H14O2 Formula Weight: 142.20 Melting Point: 109-110° MDL No.: MFCD00074943

Formel: C₈H₁₄O₂
MW: 142.2 g/mol
Melting Pt: 109…110 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00074943
CAS-Nummer: 13064-83-0
EINECS: 235-959-6

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR