4-Aminopyrimidin 98%

Lieferant: Thermo Fisher Scientific
Achtung

H27367.MD H27367.06 H27367.03
H27367.MDEA 116 CHF
H27367.MD H27367.06 H27367.03
4-Aminopyrimidin 98%
4-Aminopyrimidin

Purity: 98% CAS No.: 591-54-8 Molecular Formula: C4H5N3 Formula Weight: 95.11 Melting Point: 147-152° MDL No.: MFCD00006112

Formel: C₄H₅N₃
MW: 95.1 g/mol
Melting Pt: 147…152 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006112
CAS-Nummer: 591-54-8

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR