3-Brom-2,6-difluorbenzaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H26213.06 H26213.14
H26213.06EA 271 CHF
H26213.06 H26213.14
3-Brom-2,6-difluorbenzaldehyd ≥98%
3-Bromo-2,6-difluorobenzaldehyde

Purity: 98% CAS No.: 398456-82-1 Molecular Formula: C7H3BrF2O Formula Weight: 221.00 Melting Point: 53-57° MDL No.: MFCD08282773

Formel: C₇H₃BrF₂O
MW: 221 g/mol
Melting Pt: 53…57 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD08282773
CAS-Nummer: 398456-82-1
EINECS: None

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR