Cryoboxes B13/B15/B25
Lieferant: GLW
B25B B15 B15B B15G B15R B15Y B25 B25G B25R B25Y B25S B13 B13B B13G B13R B13S B13Y B15BS B15GS B15RS B15S B15YS B25BS B25GS B25RS B25YS BS13 BS15 BS25
GLWAB25BEA 26.2 CHF
GLWAB25B GLWAB15 GLWAB15B GLWAB15G GLWAB15R GLWAB15Y GLWAB25 GLWAB25G GLWAB25R GLWAB25Y GLWAB25S GLWAB13 GLWAB13B GLWAB13G GLWAB13R GLWAB13S GLWAB13Y GLWAB15BS GLWAB15GS GLWAB15RS GLWAB15S GLWAB15YS GLWAB25BS GLWAB25GS GLWAB25RS GLWAB25YS GLWABS13 GLWABS15 GLWABS25
Cryoboxes B13/B15/B25
Boxen Kryoboxen
PP, natural or coloured.
  • Autoclavable up to +121 °C and use in freezer down to -90 °C
  • The included clear lid offers a view of the contents without opening the cryobox
  • Drainage holes in base for easy opening the box at low temperatures
  • For light sensitive samples please choose the color Black/black
Suitable for 25 tubes, arranged in 5 x 5 compartments with 17 mm diameter
B13-Series, height variable 70-80 mm
Designed for 6 ml up to 9 ml sample vials and test tubes. Generic tube size up to 80 mm height and 17 mm diameter.
B15-Series, height variable 95-1More Product Information