3-Brom-5-formylpyridin

Synonyms: 3-Bromopyridine-5-carboxaldehyde, 5-Bromopyridine-3-carboxaldehyde, 5-Bromonicotinaldehyde, 5-Bromo-3-pyridinecarboxaldehyde

037001-5G
FLUO037001-5GEA 0 CHF
FLUO037001-5G
3-Brom-5-formylpyridin
3-Brom-5-formylpyridin
Formel: C₆H₄BrNO
MW: 186,01 g/mol
MDL Number: MFCD03265758
CAS-Nummer: 113118-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR