Restriction enzymes, XhoI, FastDigest™, Fermentas
Lieferant: Thermo Fisher Scientific
FD0694 FD0695
FD0694EA 96.8 CHF
FD0694 FD0695
Restriction enzymes, XhoI, FastDigest™, Fermentas
Nukleinsäure-Reagenzien Restriktionsenzym
FastDigest™ XhoI is one of an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest™ and FastDigest™ Green reaction buffers.
  • 100% buffer compatibility with downstream applications
  • Complete digestion in 5 to 15 minutes
  • Direct loading on gels
Order Now
TECHNISCHE DATEN
Environmentally Preferable
Anwendung
Enzymname XhoI
Ursprung
Overhang type 5' overhang
Kompatibler Puffer FastDigest Green Buffer or FastDigest Buffer (colorless)