Aqueous mounting medium, Fluoroshield™, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

F6182-20ML
F6182-20MLEA 102 CHF
F6182-20ML
Aqueous mounting medium, Fluoroshield™, Sigma-Aldrich®
Färbemittel Mikroskopie- und Färbereagenzien

Fluoroshield™ is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.


  • Storage temperature: 2 - 8 °C


This unique formula prevents rapid photobleaching of FITC, Texas Red®, AMCA, Cy®, Cy3™, Cy5, Alexa Fluor® 488, Alexa Fluor® 594, Green fluorescent protein (GFP), tetramethylrhodamine, R-phycoerythrin (R-PE), phycocyanin (PC), and allophycocyanin (APC).


Applications:
- Mounting medium for cultured megakaryocytes and immunostained platelets for confocal microscopy studies
- Mounting medium for 4′,6-diamidino-2-phenylindole (DAPI) stained heart sections for immunofluorescence studies
- Preserve fluorescence of C6 cell smears

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR