Formalinlösung 10% Histological Fixative, neutral buffered

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol

MQ200

F5554-19L F5554-4L
F5554-19LEA 243 CHF
F5554-19L F5554-4L
Formalinlösung 10% Histological Fixative, neutral buffered
Formalinlösung

10% Neutral buffered formalin (NBF) is the most commonly used fixative for light microscopy. 10% NBF is well suited for large throughput laboratories and requires a relatively short period of fixation, but can also be used for the long-term storage of tissue. Formalin fixation is thought to form between the aldehydes and the proteins, creating a gel, thus retaining cellular constituents in their in vivo relationship.


  • Formalin, 10%, Neutral Buffered, Wintergreen Scented has been used as a fixative in transformation and tumorigenicity assay[3] and in immunohistostaining
  • Intended for use as a general purpose histological fixative
  • Wintergreen oil has been added to the standard formulation to provide a wintergreen scent

Formel: CH₂O
MW: 30.03 g/mol
Lager Temperatur: Room Temperature
MDL Number: MFCD00003274
CAS-Nummer: 50-00-0

Order Now

Spezifikation Testergebnisse

Appearance (Colour) Colorless
Appearance (Form) Liquid
Appearance (Turbidity) Clear
pH 6.9 - 7.1
Assay
% Formaldehyde (w/v)
3.5 - 4.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR