Avantor® ACE®, UHPLC Pre-Column Filters
Lieferant: Avantor
EXL-PCF10/ACQEA 874 CHF
EXL-PCF10/ACQ EXL-PCF10 EXL-PCF05/ACQ EXL-PCF05 EXL-PCF01/ACQ EXL-PCF01
Avantor® ACE®, UHPLC Pre-Column Filters
Filter Vorsäulen-Filter und -Fritten
Avantor® ACE® UHPLC pre-column filters made from stainless steel/PEEK provide protection for all UHPLC columns. They offer ultra-low dispersion for maximum compatibility with UHPLC instruments and columns. For use with Waters 'Classic' Acquity® systems, an alternative Waters Acquity® compatible version provides improved performance.
  • Universal filter, suitable for all UHPLC columns
  • Compatible with all UHPLC systems
  • Pressure rated to 1000 bar
  • Reusable
Order Now
TECHNISCHE DATEN
Environmentally Preferable