Exsil Silica, HPLC Columns
Lieferant: Avantor
EX-5-250AEA 477 CHF
EX-5-250A EX-10-250P EX80-5-250A
Exsil Silica, HPLC Columns
Chromatographiesäulen
Exsil HPLC columns are a useful alternative to the classical spherical HPLC silicas.
  • Spherical porous silica
  • 80 and 100 Å pore sizes
  • Competitively priced
  • Hichrom high efficiency
Exsil 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications. Exsil 80 is a higher surface area, complementary range of materials.