Cutan® Wall Dispenser for Hand Sanitiser
Lieferant: Deb-STOKO
CPROB01SA
EVONCPROB01SAEA 12.3 CHF
EVONCPROB01SA
Cutan® Wall Dispenser for Hand Sanitiser
Seifen- und Flüssigseifenspender
Cutan dispenser for use with any Cutan 1 Litre hand sanitiser cartridge.
  • Contains silver phosphate glass technology to prevent microbial growth
  • Strong and durable
  • Key operated lock prevents theft
  • Large push button that can be hand or elbow operated
  • Easy to slot in and replace the cartridge
Red coloured label and push button for easy distinction and identification of hand sanitiser amongst hand cleaning and moisturising agents. No special DIY skills required, can be easily mounted with the sticky pad at the back of the dispenser. Unit self-primes onMore Product Information
New Product
Order Now
TECHNISCHE DATEN
Environmentally Preferable