Hand cleansing creams, Kresto® paint liquid
Lieferant: Deb-STOKO
22306
EVON22306EA 341 CHF
EVON22306
Hand cleansing creams, Kresto® paint liquid
Hautpflege Seifen und Hautreiniger
The special hand cleanser with very powerful cleansing action contains mild solvents. It removes very stubborn and extremely heavy-duty contamination like lacquers, resins, adhesives, oil-based paints.
  • Scrubbing agent-free
  • As the formulation is concentrated, only a small amount is required to remove contaminants
  • Solvent-free, colourant-free, preservative-free, perfumed
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904
Order Now
TECHNISCHE DATEN
Environmentally Preferable