Anti-IDE Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB07899
EVBIEB07899EA 395 CHF
EVBIEB07899
Anti-IDE Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IDE (insulin-degrading enzyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR