Lithiumacetat Dihydrat

Lieferant: EUROCLONE

Synonyms: Acetic acid lithium salt

EMR162001 EMR162100 EMR162500
EURLEMR162001EA 0 CHF
EURLEMR162001 EURLEMR162100 EURLEMR162500
Lithiumacetat Dihydrat
Lithiumacetat Dihydrat
Formel: H₃CCOOLi·2H₂O
Melting Pt: 291 °C
Dichte: 1.26 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00066949
CAS-Nummer: 6108-17-4
EINECS: 208-914-3
Merck Index: 13,05543

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR