InVue® NT6520 Integrated Flow Controllers
Lieferant: ENTEGRIS
6520T7F4B12KP7 6520T8F6B12AP7 6520T8F6B12KP7
ENTE6520T7F4B12KP7EA 0 CHF
ENTE6520T7F4B12KP7 ENTE6520T8F6B12AP7 ENTE6520T8F6B12KP7
InVue® NT6520 Integrated Flow Controllers
Durchflussregler
Offers precise point-of-use chemical flow control and is designed for point-of-use blending and dispense applications requiring medium to high flow rates (2,5 to 40 L/min).
  • Most accurate in bubble prone media vs. other commonly used technologies
  • Integral pressure transducer for simultaneous dual output - flow and pressure
  • Compact footprint for space savings and easy installation
  • High repeatability for superior process control
The InVue Integrated Flow Controller, NT6520 (2,5 to 40 L/min) combines our differential pressure based flowmeter and leading-More Product Information