Water purification systems for healthcare applications, BIOPURE 7/15
Lieferant: ELGA LabWater
MB007BPM1 MB007PK01 MB007XXM1 MB015BPM1 MB015PK01
ELGAMB007BPM1EA 0 CHF
ELGAMB007BPM1 ELGAMB007PK01 ELGAMB007XXM1 ELGAMB015BPM1 ELGAMB015PK01
Water purification systems for healthcare applications, BIOPURE 7/15
Wasseraufbereitungssysteme Reinstwassersysteme
These systems are designed to deliver compliant purified water at the point of use for a wide range of washer disinfectors and autoclaves to minimise contamination risk to patients.
  • Cost effective: Provides the convenience of on-demand pure water at a price comparable with purified bottled water
  • Space saving: Compact size means that it can be positioned under a bench, wall mounted or fitted into a cabinet
  • Guaranteed bacterial specification, provided by integrated purification technologies combined with easy system disinfection
  • Water specification is
More Product Information