3-Chlorbiphenyl (PCB Nr. 2)

Lieferant: EHRENSTORFER

C20000200
EHERC20000200EA 53.4 CHF
EHERC20000200
3-Chlorbiphenyl (PCB Nr. 2)
3-Chlorbiphenyl (PCB Nr. 2)
Formel: C₁₂H₉Cl
Siedepunkt: 285 °C (1013 hPa)
Melting Pt: 16 °C
Dichte: 1.1579 g/cm³ (25 °C)
MDL Number: MFCD00013633
CAS-Nummer: 2051-61-8
EINECS: 218-126-1
UN: 2315
ADR: 9,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR