2,3,5-Triiodbenzoesäure

Lieferant: EHRENSTORFER
Achtung

Synonyms: TIBA

C17870000
EHERC17870000EA 107 CHF
EHERC17870000
2,3,5-Triiodbenzoesäure
2,3,5-Triiodbenzoesäure
Formel: C₇H₃I₃O₂
MW: 499.81 g/mol
Siedepunkt: 416
Melting Pt: 225,9
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00002420
CAS-Nummer: 88-82-4
EINECS: 201-859-6

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR