n-Tetradecan

Lieferant: EHRENSTORFER
Gefahr

Synonyms: Tetradecane

C17396500
EHERC17396500EA 68.9 CHF
EHERC17396500
n-Tetradecan
n-Tetradecan
Formel: H₃C(CH₂)₁₂CH₃
Siedepunkt: 253.7 °C (1013 hPa)
Melting Pt: 6 °C
Dichte: 0.7628 g/cm³ (20 °C)
Flash-Pt: 102 °C (closed cup)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008986
CAS-Nummer: 629-59-4
EINECS: 211-096-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03064a29015d1e810f1e4350a7a6cd2dfe4713fd&vid=f4838a5686fe033622495a3bf280963555654d9a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR