2,4,5-T-Butylester (2,4,5-Trichlorphenoxyessigsäurebutylester)

Lieferant: EHRENSTORFER

Synonyms: Butyl (2,4,5-trichlorophenoxy)acetate

C17105500
EHERC17105500EA 195 CHF
EHERC17105500
2,4,5-T-Butylester (2,4,5-Trichlorphenoxyessigsäurebutylester)
2,4,5-T-Butylester (2,4,5-Trichlorphenoxyessigsäurebutylester)
Formel: C₁₂H₁₃Cl₃O₃
MDL Number: MFCD00128016
CAS-Nummer: 93-79-8
EINECS: 202-277-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR