Phenylbutazon

Lieferant: EHRENSTORFER

Synonyms: 4-Butyl-1,2-diphenylpyrazolidine-3,5-dione

C16056500
EHERC16056500EA 82.7 CHF
EHERC16056500
Phenylbutazon
Phenylbutazon
Formel: C₁₉H₂₀N₂O₂
MDL Number: MFCD00005500
CAS-Nummer: 50-33-9
EINECS: 200-029-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR