(2-Naphthoxy)essigsäure

Lieferant: EHRENSTORFER

Synonyms: 2-Naphthoxyacetic acid

C15439000
EHERC15439000EA 75.8 CHF
EHERC15439000
(2-Naphthoxy)essigsäure
(2-Naphthoxy)essigsäure
Formel: C₁₂H₁₀O₃
MW: 202,21 g/mol
MDL Number: MFCD00004066
CAS-Nummer: 120-23-0
EINECS: 204-380-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR