6-Ethyl-o-toluidin

Lieferant: EHRENSTORFER

Synonyms: 2-Ethyl-6-methylaniline, 2-Amino-3-ethyltoluene

C13348000
EHERC13348000EA 74 CHF
EHERC13348000
6-Ethyl-o-toluidin
6-Ethyl-o-toluidin
Formel: C₉H₁₃N
MW: 135,21 g/mol
Siedepunkt: 231 °C (1013 hPa)
Melting Pt: –33 °C
Dichte: 0,969 g/cm³ (20 °C)
MDL Number: MFCD00007752
CAS-Nummer: 24549-06-2
EINECS: 246-309-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR