Dopamin Hydrochlorid

Lieferant: EHRENSTORFER

Synonyms: 1-(2-Aminoethyl)-3,4-dihydroxybenzene HCl, 3-Hydroxytyraminium chloride, 3-Hydroxytyramine hydrochloride, 3,4-Dihydroxyphenethylammonium chloride

C13082000
EHERC13082000EA 149 CHF
EHERC13082000
Dopamin Hydrochlorid
Dopamin Hydrochlorid
Formel: C₈H₁₂ClNO₂
Melting Pt: 246…249 °C
Dichte: 1,356 g/cm³ (20 °C)
MDL Number: MFCD00012898
CAS-Nummer: 62-31-7
EINECS: 200-527-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR