Dinoseb (2-sec-butyl-4,6-dinitrophenol)

Lieferant: EHRENSTORFER

C12810000
EHERC12810000EA 133 CHF
EHERC12810000
Dinoseb (2-sec-butyl-4,6-dinitrophenol)
Dinoseb (2-sec-Butyl-4,6-dinitrophenol)
Formel: C₁₀H₁₂N₂O₅
MW: 240,22 g/mol
CAS-Nummer: 88-85-7
EINECS: 201-861-7
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR