2-Chlor-5-(chlormethyl)pyridin

Lieferant: EHRENSTORFER

C11397000
EHERC11397000EA 143 CHF
EHERC11397000
2-Chlor-5-(chlormethyl)pyridin
2-Chloro-5-(chloromethyl)pyridine
Formel: C₆H₅Cl₂N
Melting Pt: 37 °C
MDL Number: MFCD00125366
CAS-Nummer: 70258-18-3
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR