Clamping rings for small flanges
Lieferant: EDWARDS HIGH VACUUM METTLER
C105-14-401
EDWAC105-14-401EA 0 CHF
EDWAC105-14-401
Clamping rings for small flanges
Klemmen Klemmen für den allgemeinen Gebrauch
Clamping rings made of stainless steel, for connections with NW flanges.