Ready-to-Load™ kits, what is PCR and how does it work?
Lieferant: EDVOTEK
S-48
EDVOS-48EA 71.5 CHF
EDVOS-48
Ready-to-Load™ kits, what is PCR and how does it work?
Lehrmittel Biologie-Lehrmittel Lehrmittel zur PCR
This simulation experiment demonstrates the process of DNA amplification by PCR and how the amplified product is detected by separating the reaction mixture by agarose gel electrophoresis.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
Lieferumfang: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.