VWR®, pH standard kit
Lieferant: VWR Chemicals
E603-KITEA 299 CHF
E603-KIT
VWR®, pH standard kit
Puffer pH-Puffer
Kit contains 500 ml of pH 4,0, pH 7,0 and pH 10,0 standards.
Order Now
TECHNISCHE DATEN
Environmentally Preferable