Instant hand sanitisers, SURE®
Lieferant: Diversey
101104071 100977079
DIVY101104071EA 248 CHF
DIVY101104071 115-3080
Instant hand sanitisers, SURE®
Hautpflege Desinfektionsmittel für die Haut Desinfektionsfluid und -schaum für die Haut
Disinfects hands in one easy step without the need for water. Non classified highly efficacious alcohol-free foam hand rub with a refreshing fragrance.
  • Decomposed by natural processes
  • Passes EN test standard EN 1500, EN 13727, EN 1276, EN 13624 (yeast), EN 1650 (yeast), EN 14476 (Modified Vaccinia Ankara)
  • Passes the latest BPR test requirements for hand disinfection
  • 100% natural perfume for a refreshing fragrance
  • Extensively tested skin mildness (patch test)
  • No environmental hazard classifications, no safety warnings for user
SURE® instant handMore Product Information