Dual Stage Gas Regulators
Lieferant: DISTEK
5000-2009 5000-2010 5000-2011 5000-2012
DISI5000-2009EA 0 CHF
DISI5000-2009 DISI5000-2010 DISI5000-2011 DISI5000-2012
Dual Stage Gas Regulators
Druckminderer
Dual stage gas regulator with outlet connection - ¹/₄" tube fitting.
  • Delivery range - 0 to 3,44 bar
Order Now
TECHNISCHE DATEN
Environmentally Preferable